KEY465195893 Key Organics / BIONET

N-(2,4-dinitrophenyl)-12-oxa-16-azatetracyclo[11.2.1.0^{2,11}.0^{3,8}]hexadeca-2(11),3(8),4,6,9-pentaen-16-amine

  • MDL number MFCD00173608
  • Cas Number : 22794-77-0
  • Molecular Weight : 392.371

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74836

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-020_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart