KEY465195899 Key Organics / BIONET

ethyl 2-[(1-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]amino}-2-methylpropan-2-yl)sulfanyl]acetate

  • MDL number MFCD01315023
  • Cas Number : 478042-69-2
  • Molecular Weight : 370.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74848

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-032_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart