KEY465195905 Key Organics / BIONET

5,6-dimethyl-4-nitro-1-(propan-2-yl)-1H-1,3-benzodiazole

  • MDL number MFCD00231515
  • Cas Number : 338423-01-1
  • Molecular Weight : 233.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74875

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-316S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart