KEY465195909 Key Organics / BIONET

N-[5,6-dimethyl-1-(propan-2-yl)-1H-1,3-benzodiazol-4-yl]-4-fluorobenzene-1-sulfonamide

  • MDL number MFCD00231528
  • Cas Number : 338423-30-6
  • Molecular Weight : 361.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74907

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-353S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart