KEY465195913 Key Organics / BIONET

(2Z)-2-(2,4-dichlorophenyl)-3-(dimethylamino)prop-2-enal

  • MDL number MFCD04125371
  • Cas Number : 103824-10-8
  • Molecular Weight : 244.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74922

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-368S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart