KEY465195914 Key Organics / BIONET

methyl 2-[(4,5-dimethoxypyrimidin-2-yl)sulfanyl]acetate

  • MDL number MFCD00215637
  • Cas Number : 338423-43-1
  • Molecular Weight : 244.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74923

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-369S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart