KEY465195915 Key Organics / BIONET

ethyl 2-{[5-methoxy-4-(3-nitrophenoxy)pyrimidin-2-yl]sulfanyl}acetate

  • MDL number MFCD00215638
  • Cas Number : 338423-44-2
  • Molecular Weight : 365.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74925

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-371S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart