KEY465195917 Key Organics / BIONET

6-chloro-N-phenyl-[1,2,4]triazolo[4,3-b]pyridazin-3-amine

  • MDL number MFCD04125373
  • Cas Number : 182348-50-1
  • Molecular Weight : 245.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74931

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-377S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart