KEY465195920 Key Organics / BIONET

5-(4-chlorophenyl)-1-[(2,4-dichlorophenyl)methyl]-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00231546
  • Cas Number : 338423-56-6
  • Molecular Weight : 389.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74938

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-385S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart