KEY465195921 Key Organics / BIONET

5-(4-chlorophenyl)-1-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00231547
  • Cas Number : 338423-57-7
  • Molecular Weight : 244.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74939

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-386S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart