KEY465195924 Key Organics / BIONET

ethyl 2-({4-[(4-chlorophenyl)sulfanyl]-5-methoxypyrimidin-2-yl}sulfanyl)propanoate

  • MDL number MFCD00215642
  • Cas Number : 338423-63-5
  • Molecular Weight : 384.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74947

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-394S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart