KEY465195926 Key Organics / BIONET

1-[(4-methoxyphenyl)methyl]-5,6-dimethyl-1H-1,3-benzodiazol-4-amine

  • MDL number MFCD00231554
  • Cas Number : 338423-66-8
  • Molecular Weight : 281.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/74951

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4J-398S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart