KEY465195936 Key Organics / BIONET

N-(4-chlorophenyl)-2-hydroxy-3-(4-methoxybenzenesulfonyl)-2-methylpropanamide

  • MDL number MFCD00231753
  • Cas Number : 338424-26-3
  • Molecular Weight : 383.84

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75000

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-546S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart