KEY465195940 Key Organics / BIONET

3-(benzenesulfonyl)-N-(2,4-dichlorophenyl)-2-hydroxy-2-methylpropanamide

  • MDL number MFCD00232088
  • Cas Number : 339275-79-5
  • Molecular Weight : 388.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75037

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4J-587S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart