KEY465195956 Key Organics / BIONET

3,3-bis(methylsulfanyl)-2-(4-nitrophenyl)prop-2-enenitrile

  • MDL number MFCD00111968
  • Cas Number : 29866-34-0
  • Molecular Weight : 266.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75129

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4K-002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart