KEY465195958 Key Organics / BIONET

(2E,4Z)-4-(benzenesulfonyl)-5-(dimethylamino)penta-2,4-dienenitrile

  • MDL number MFCD00975562
  • Cas Number : 339276-59-4
  • Molecular Weight : 262.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75131

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4K-005_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart