KEY465195959 Key Organics / BIONET

2-{[(2E)-3-cyanoprop-2-en-1-yl]sulfanyl}-4,6-dimethylpyridine-3-carbonitrile

  • MDL number MFCD00975563
  • Cas Number : 339276-61-8
  • Molecular Weight : 229.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75136

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4K-011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart