KEY465195960 Key Organics / BIONET

(2E,4E)-4-(benzenesulfonyl)-5-{[4-(trifluoromethyl)phenyl]amino}penta-2,4-dienenitrile

  • MDL number MFCD00975565
  • Cas Number : 339276-63-0
  • Molecular Weight : 378.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75138

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4K-013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart