KEY465195967 Key Organics / BIONET

2-(3,4-dichlorophenyl)-1-phenylmethanesulfonylpropan-2-ol

  • MDL number MFCD00244875
  • Cas Number : 339276-78-7
  • Molecular Weight : 359.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75161

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4K-308S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart