KEY465195978 Key Organics / BIONET

1-(2,4-dichlorophenyl)-6-methyl-2-oxo-4-(trifluoromethyl)-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD01314970
  • Cas Number : 339276-98-1
  • Molecular Weight : 347.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75203

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4K-359S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart