KEY465195986 Key Organics / BIONET

2-methyl-3-phenyl-octahydro-1,2-benzoxazol-4-one

  • MDL number MFCD00974227
  • Cas Number : 318248-52-1
  • Molecular Weight : 231.295

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75337

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4K-903_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart