KEY465195992 Key Organics / BIONET

ethyl N-[(2Z)-3-oxo-2-{2-[4-(trifluoromethyl)phenyl]hydrazin-1-ylidene}butanoyl]carbamate

  • MDL number MFCD00794263
  • Cas Number : 478042-73-8
  • Molecular Weight : 345.278

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75355

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4K-928_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart