KEY465195993 Key Organics / BIONET

6-acetyl-2-[4-(trifluoromethyl)phenyl]-2,3,4,5-tetrahydro-1,2,4-triazine-3,5-dione

  • MDL number MFCD00794264
  • Cas Number : 478042-74-9
  • Molecular Weight : 299.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75356

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4K-929_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart