KEY465195995 Key Organics / BIONET

5-methyl-N-[2-(pyridin-2-yl)ethyl]thieno[2,3-d]pyrimidin-4-amine

  • MDL number MFCD00096435
  • Cas Number : 439108-19-7
  • Molecular Weight : 270.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75364

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4L-007_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart