KEY465196004 Key Organics / BIONET

(3E)-3-(methoxyimino)-2-(3-oxo-1,3-dihydro-2-benzofuran-1-yl)propanenitrile

  • MDL number MFCD00663702
  • Cas Number : 339278-31-8
  • Molecular Weight : 230.223

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75384

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-031_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart