KEY465196005 Key Organics / BIONET

(2Z)-2-[(1E)-(methoxyimino)methyl]-3-(thiophen-2-yl)prop-2-enenitrile

  • MDL number MFCD00663703
  • Cas Number : 339278-32-9
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75385

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-032_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart