KEY465196007 Key Organics / BIONET

(1E,4E)-1-(2,4-dichlorophenyl)-5-(dimethylamino)penta-1,4-dien-3-one

  • MDL number MFCD00244365
  • Cas Number : 339278-34-1
  • Molecular Weight : 270.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75388

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4L-035_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart