KEY465196011 Key Organics / BIONET

3-(4-chlorophenyl)-1-[(E)-2-phenylethenyl]urea

  • MDL number MFCD00794892
  • Cas Number : 1164507-93-0
  • Molecular Weight : 272.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-308S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart