KEY465196021 Key Organics / BIONET

6-{[(4-chlorophenyl)sulfanyl]methyl}-N-[(3-methoxyphenyl)methyl]-2-methylpyrimidin-4-amine

  • MDL number MFCD00794929
  • Cas Number : 339278-87-4
  • Molecular Weight : 385.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75465

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
MSDS https://www.keyorganics.net/media/catalog/product/sds/4L-394S_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart