KEY465196025 Key Organics / BIONET

ethyl 2-[3-cyano-4-(methylsulfanyl)-2-oxo-6-phenyl-1,2-dihydropyridin-1-yl]acetate

  • MDL number MFCD00794502
  • Cas Number : 478042-84-1
  • Molecular Weight : 328.39

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75491

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-537S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart