KEY465196027 Key Organics / BIONET

6-(methoxymethyl)-2-(pyridin-3-yl)pyrimidin-4-ol

  • MDL number MFCD00268711
  • Cas Number : 339278-99-8
  • Molecular Weight : 217.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75501

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-555S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart