KEY465196028 Key Organics / BIONET

6-(methoxymethyl)-2-phenyl-N-(propan-2-yl)pyrimidin-4-amine

  • MDL number MFCD00794782
  • Cas Number : 339279-03-7
  • Molecular Weight : 257.337

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75505

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-559S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart