KEY465196031 Key Organics / BIONET

6-(4-chlorophenyl)-1-[(2-methylphenyl)methyl]-4-(methylsulfanyl)-2-oxo-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00794511
  • Cas Number : 478042-92-1
  • Molecular Weight : 380.89

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75519

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4L-575S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart