KEY465196037 Key Organics / BIONET

1-[(3-methylphenyl)methyl]-4-(methylsulfanyl)-2-oxo-6-phenyl-1,2-dihydropyridine-3-carbonitrile

  • MDL number MFCD00794520
  • Cas Number : 478043-01-5
  • Molecular Weight : 346.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75528

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-584S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart