KEY465196040 Key Organics / BIONET

5-[(5-cyano-3-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)methyl]-4,5-dihydro-1,2-oxazole-3-carbohydrazide

  • MDL number MFCD01315192
  • Cas Number : 339279-29-7
  • Molecular Weight : 292.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75550

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-902_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart