KEY465196041 Key Organics / BIONET

ethyl (3Z)-3-({2-[(E)-(4-ethoxy-4-oxobutan-2-ylidene)amino]ethyl}imino)butanoate hydrate

  • MDL number MFCD26383447
  • Cas Number : 1798415-80-1
  • Molecular Weight : 302.371

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75553

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4L-905_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart