KEY465196043 Key Organics / BIONET

ethyl 2-({[2-hydroxy-2-(naphthalen-2-yl)ethyl]carbamothioyl}amino)acetate

  • MDL number MFCD00794664
  • Cas Number : 339279-36-6
  • Molecular Weight : 332.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75560

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4L-913_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart