KEY465196045 Key Organics / BIONET

(2E)-3-oxo-3-phenyl-2-{2-[3-(trifluoromethyl)phenyl]hydrazin-1-ylidene}propanal

  • MDL number MFCD00975129
  • Cas Number : 339279-39-9
  • Molecular Weight : 320.271

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75563

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-002_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart