KEY465196048 Key Organics / BIONET

(2E)-2-[2-(4-methoxyphenyl)hydrazin-1-ylidene]-3-oxo-3-phenylpropanal

  • MDL number MFCD00975132
  • Cas Number : 77083-67-1
  • Molecular Weight : 282.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75572

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart