KEY465196051 Key Organics / BIONET

2-phenyl-5-[1-(thiophene-2-sulfonyl)-1H-pyrazol-3-yl]-1,3-thiazole

  • MDL number MFCD00974316
  • Cas Number : 318248-79-2
  • Molecular Weight : 373.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75582

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-021_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart