KEY465196054 Key Organics / BIONET

5-[(Z)-2-(2-chlorophenyl)diazen-1-yl]-4-phenylpyrimidin-2-amine

  • MDL number MFCD00974797
  • Cas Number : 339279-61-7
  • Molecular Weight : 309.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75598

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4M-037_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart