KEY465196060 Key Organics / BIONET

(Z)-N-[3,3-dimethyl-1-(2-nitrophenoxy)butan-2-ylidene]hydroxylamine

  • MDL number MFCD02083110
  • Cas Number : 478043-05-9
  • Molecular Weight : 252.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75631

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-311S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart