KEY465196063 Key Organics / BIONET

4-(4-chlorobenzenesulfonyl)-3-methyl-3,4-dihydro-2H-1,4-benzoxazine

  • MDL number MFCD01568988
  • Cas Number : 338747-51-6
  • Molecular Weight : 323.79

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75640

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-324S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart