KEY465196064 Key Organics / BIONET

(E)-[3,3-dimethyl-1-(2-nitrophenoxy)butan-2-ylidene]amino N-(2,6-dimethylphenyl)carbamate

  • MDL number MFCD02083116
  • Cas Number : 478043-12-8
  • Molecular Weight : 399.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75651

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4M-338S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart