KEY465196070 Key Organics / BIONET

methyl 4-[(2,3-dichlorophenyl)sulfanyl]pyridine-2-carboxylate

  • MDL number MFCD01444073
  • Cas Number : 338748-09-7
  • Molecular Weight : 314.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75702

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4M-562S_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart