KEY465196074 Key Organics / BIONET

ethyl 2-methyl-5-oxo-4-phenyl-5,6,7,8-tetrahydroquinoline-3-carboxylate

  • MDL number MFCD00974811
  • Cas Number : 154405-24-0
  • Molecular Weight : 309.365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75720

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-717_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart