KEY465196077 Key Organics / BIONET

5,6,14,14-tetramethyl-10-[4-(propan-2-yl)phenyl]-2,9-diazatricyclo[9.4.0.0^{3,8}]pentadeca-1(11),3,5,7-tetraen-12-one

  • MDL number MFCD00794579
  • Cas Number : 338748-43-9
  • Molecular Weight : 388.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75732

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-735_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart