KEY465196089 Key Organics / BIONET

2,3-dimethyl 1-[(4-methoxyphenyl)methyl]-6-oxo-1,4,5,6-tetrahydropyridine-2,3-dicarboxylate

  • MDL number MFCD00975044
  • Cas Number : 338748-78-0
  • Molecular Weight : 333.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75778

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-931_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart