KEY465196092 Key Organics / BIONET

ethyl 1-{2-[5-(ethoxycarbonyl)-6-methyl-2-oxo-1,2,3,4-tetrahydropyridin-1-yl]ethyl}-2-methyl-6-oxo-1,4,5,6-tetrahydropyridine-3-carboxylate

  • MDL number MFCD00975056
  • Cas Number : 338748-90-6
  • Molecular Weight : 392.452

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Storage temp. RT
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75792

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/4M-948_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart