KEY465196096 Key Organics / BIONET

3-[(1E)-(dimethylamino)methylidene]-1-(5-iodopyridin-2-yl)thiourea

  • MDL number MFCD01315200
  • Cas Number : 338748-97-3
  • Molecular Weight : 334.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/75800

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/4M-959_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart